VWR®, Bovine serum albumin (BSA), Cohn fraction V
Leverandør: VWR Chemicals
422371XEA 5400 NOK
422371X
VWR®, Bovine serum albumin (BSA), Cohn fraction V
Proteiner
A Cohn fraction V albumin produced by cold ethanol fractionation with minimum use of chemicals. Fatty acids and native sialic acid residues are preserved in this BSA.