VWR®, Bovine serum albumin (BSA)
Leverandør: VWR Chemicals
422381BEA 8525 NOK
422381B
VWR®, Bovine serum albumin (BSA)
Proteiner
Produced by a heat shock process using sodium caprylate as a stabiliser with additional treatments including dialysis and deionisation.
  • Low endotoxin