Wolfram 100000 mg/L i vann (fra (NH₄)₂WO₄) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

42249.AD 42249.AE 42249.AP
42249.ADEA 2075 NOK
42249.AD 42249.AE 42249.AP
Wolfram 100000 mg/L i vann (fra (NH₄)₂WO₄) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Tungsten Standards

Molecular Formula: (NH₄)₂WO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00011461


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011461

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR