Stirring Rods, Thermo Fisher Scientific
Leverandør: Thermo Fisher Scientific
42294.NK 42293.NK 42292.NT 42293.NT 42294.NT
42294.NKEA 425 NOK
42294.NK 42293.NK 42292.NT 42293.NT 42294.NT
Stirring Rods, Thermo Fisher Scientific
Omrørere Omrørerstaver
The fused quartz rods are manufactured by fusing naturally occurring crystalline silica.
  • Fused quartz has high chemical purity, high corrosion resistance and high melting point
  • Quartz is extremely hard, with a low coefficient of thermal expansion, excellent electrical insulation qualities and optical transmission from ultra-violet to infra-red