Antimony standard dissolved in oil, 5.000 µg/g Sb in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

42433.1Y 42433.18
42433.1YEA 920 NOK
42433.1Y 42433.18
Antimony standard dissolved in oil, 5.000 µg/g Sb in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Antimony Standards

Physical form: liquid
MDL No.: MFCD00134030

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR