Krom standard dissolved in oil, 5.000 µg/g Cr in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

42438.1Y 42438.18
42438.1YEA 920 NOK
42438.1Y 42438.18
Krom standard dissolved in oil, 5.000 µg/g Cr in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Chromium Standards

Physical Form: Liquid
MDL No.: MFCD00010944

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR