Iron standard dissolved in oil, 5.000 µg/g Fe in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

42440.18
42440.18EA 1575 NOK
42440.18
Iron standard dissolved in oil, 5.000 µg/g Fe in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Iron Standards

Physical form: liquid
MDL No.: MFCD00010999

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR