Zinc standard dissolved in oil, 5.000 µg/g Zn in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

42453.18
42453.18EA 1875 NOK
42453.18
Zinc standard dissolved in oil, 5.000 µg/g Zn in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Zinc Standards

Physical form: liquid
MDL No.: MFCD00011291

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR