ICP-MS stock standard solution A for 200,8, Rev. 5,4, Specpure®

Leverandør: Thermo Fisher Scientific

42596.AEEA 5325 NOK
42596.AE
ICP-MS stock standard solution A for 200,8, Rev. 5,4, Specpure®
Standarder Kationstandarder Multi-Element Standards

UN No.: UN3264
MDL No.: MFCD03792352

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD03792352

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR