ICP-MS analytes B for 6020 CLP-M, Specpure®

Leverandør: Thermo Fisher Scientific

42650.AEEA 2200 NOK
42650.AE
ICP-MS analytes B for 6020 CLP-M, Specpure®
Standarder Kationstandarder Multi-Element Standards

11 elements


UN No.: UN3264
MDL No.: MFCD02262473


Concentration:
Ag 5 µg/ml ±5%
As 10 µg/ml ±2%
Cd 10 µg/ml ±2%
Co 20 µg/ml ±2%
Cr 20 µg/ml ±2%
Cu 20 µg/ml ±2%
Mn 20 µg/ml ±2%
Ni 20 µg/ml ±2%
Se 10 µg/ml ±2%
V 20 µg/ml ±2%
Zn 10 µg/ml ±2%

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR