Zink ≥99.95% (metals basis), metalltråd, Ø 0.25 mm (0.01 in)

Leverandør: Thermo Fisher Scientific

42703.G5 42703.G9
42703.G5EA 1875 NOK
42703.G5 42703.G9
Zink ≥99.95% (metals basis), metalltråd, Ø 0.25 mm (0.01 in)
Zinc

Purity: 99.95% CAS No.: 7440-66-6 Physical Form: ≈0.35g/m Odour: Odourless MDL No.: MFCD00011291

Formel: Zn
Kokepunkt: 908 °C (1013 hPa)
Smeltepunkt: 420 °C
Tetthet: 7,14 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011291
CAS-nummer: 7440-66-6
EINECS: 231-175-3
Merck Indeks: 12,10255

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR