Pivalaldehyde (trimethylacetaldehyde) 96%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

427201000
427201000.EA 6875 NOK
427201000.
Pivalaldehyde (trimethylacetaldehyde) 96%, AcroSeal®
Pivalaldehyde (trimethylacetaldehyde)
Formel: C₅H₁₀O
MW: 86.13 g/mol
Kokepunkt: 74
Smeltepunkt: 6
Tetthet: 0.775 g/cm³
Flash punkt: –15 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006962
CAS-nummer: 630-19-3
FN: 1989
ADR: 3 II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3800 (20°C, 589 nm)
GC ≥95 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR