Diphenylphosphine 95%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Danger

427290500
427290500.EA 5025 NOK
427290500.
Diphenylphosphine 95%, AcroSeal®
Diphenylphosphine
Formel: C₁₂H₁₁P
Tetthet: 1,07 g/cm³
Flash punkt: 110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003040
CAS-nummer: 829-85-6
FN: 2845
ADR: 4.2,I

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6250 to 1.6300 (20°C, 589 nm)
GC ≥94 %
UV at 250 nm A: ≤0.01
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0001 meq/g
Gradient analysis :
Absorbance largest eluted peak ≤0.002 A.U.
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR