Alizarin red S (sodium salt) 1% (w/v) in aqueous solution

Leverandør: Thermo Fisher Scientific

Synonymer: C.I. 58005

42746.AE 42746.AP
42746.AEEA 510 NOK
42746.AE 42746.AP
Alizarin red S (sodium salt) 1% (w/v) in aqueous solution
Alizarin red S (sodium salt)

CAS No.: 130-22-3 Molecular Formula: C14H7O7SNa Formula Weight: 342.27 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00013049

Formel: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR