Cresol red sodium salt 0,04% (w/v) in aqueous solution

Leverandør: Thermo Fisher Scientific

Synonymer: Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate

42752.AEEA 410 NOK
42752.AE
Cresol red sodium salt 0,04% (w/v) in aqueous solution
Cresol red sodium salt

CAS No.: 62625-29-0 Molecular Formula: C21H17O5S Formula Weight: 404.42 Physical Form: Liquid MDL No.: MFCD00001618

Formel: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001618
CAS-nummer: 62625-29-0
EINECS: 263-654-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR