Multi-element plasma standard solution 4, Specpure®

Leverandør: Thermo Fisher Scientific

42885.AEEA 6625 NOK
42885.AE
Multi-element plasma standard solution 4, Specpure®
Standarder Kationstandarder Multi-Element Standards

UN No.: UN3264
MDL No.: MFCD00212340

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR