Grafitt ≥99.8% (metals basis), folie, Thickness: 0.13 mm (0.005 in)

Leverandør: Thermo Fisher Scientific

43078.CJ 43078.RF 43078.RX
43078.CJEA 2475 NOK
43078.CJ 43078.RF 43078.RX
Grafitt ≥99.8% (metals basis), folie, Thickness: 0.13 mm (0.005 in)
Grafitt

Purity: 99.8% MDL No.: MFCD00144065

Formel: C
Smeltepunkt: 3827 °C
Tetthet: 2,09…2.23 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00144065
CAS-nummer: 7782-42-5
EINECS: 231-955-3
Merck Indeks: 12,04560

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR