Homogeniser, POLYTRON®, PT 6100 D
Leverandør: KINEMATICA
11010110 11010112 11010111
431-0194EA 0 NOK
431-0194 431-0219 431-0220
Homogeniser, POLYTRON®, PT 6100 D
Homogenisatorer Homogenisatorer, bordmodell
Stand model, volume range 0,1 ml to 30 litres. The powerful PT 6100D was designed to deliver exactly reproducible homognising conditions for volumes up to 30 litres with the option to select and vary speed and time of operation, whilst monitoring the temperature. The unit can be controlled via the membrane keypad or from a PC using KIN software via an RS232 interface.
  • More than 30 dispersing tools/aggregates with different geometries, designs, sizes and materials are available for selection
  • High tip speed, up to 32 m/s, for efficient