Homogeniser, high-throughput, HOHTDG
Leverandør: OHAUS
30391396
431-0261EA 0 NOK
431-0261
Homogeniser, high-throughput, HOHTDG
Homogenisatorer Homogenisatorer, bordmodell
The HT (High Throughput) lysing bead mill laboratory homogeniser provides efficient solutions for grinding, lysing, pulverising, mixing and homogenising in sample preparation applications.
  • Made with a polyurethane elastomer top housing, cycoloy base and a lexan lid
  • Flexibility to handle multiple samples and tube configurations
  • Five pre-set programs optimised for speed and time
  • Increased efficiency with high-throughput sample processing
  • Easy-to-read LED display allows the operator to view speed and time settings
  • Microprocessor-controlled unit includes