Universal mill, MultiDrive basic M20 Successor package
Leverandør: IKA
10010965
431-0308EA 60600 NOK
431-0308
Universal mill, MultiDrive basic M20 Successor package
Kverner
Regardless of whether samples are hard, soft or fibrous – IKA MultiDrive basic performs a wide range of crushing tasks. The perfect successor of the M 20 batch mill.
  • High performance - 1000 W output and variable rotational speeds ranging from 3000 rpm to 20000 rpm guarantee excellent crushing results
  • Interval operation - an asset during the coarse crushing of hard samples or for extra thorough blending
  • Integrated cooling - a cooling system is integrated in the milling cup, which allows indirect heat dissipation (in addition, dry ice can be used for cooling)