Disperser system, ULTRA-TURRAX® tube drive P control
Leverandør: IKA
25005981
431-0317EA 22100 NOK
431-0317
Disperser system, ULTRA-TURRAX® tube drive P control
Homogenisatorer Homogenisatorer, bordmodell
Universal disposable disperser system with disposable sample tubes that can be hermetically sealed. Provides protection and security from infectious sample materials, toxic substances or substances with strong odours.
  • Sealable disposable tubes optimised for your application
  • Tubes reduce the amount of cleaning required and rule out any risk of cross-contamination
  • Programmable test conditions
  • Adjustable change in direction of rotation
  • Turbo button
Programmable test conditions - user can specify and systematically program the test conditions and storeMore Product Information