Kaliumhydrogenkarbonat ≥99.95% (99.7-100.5% dry basis)

Leverandør: Honeywell Chemicals

Synonymer: Potassium bicarbonate

431583-250G 431583-50G
431583-250GEA 6025 NOK
431583-250G 431583-50G
Kaliumhydrogenkarbonat ≥99.95% (99.7-100.5% dry basis)
Kaliumhydrogenkarbonat
Formel: KHCO₃
MW: 100.12 g/mol
Smeltepunkt: 100…120 °C (dec.)
Tetthet: 2.17 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011402
CAS-nummer: 298-14-6
EINECS: 206-059-0
Merck Indeks: 13,07691

Bestill nå

Spesifikasjon Testresultater

Identity (IR) complying
Assay (excluding dried substances) 99.7 - 100.5 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 5.0 ppm
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.03 %
Metal trace analysis (ICP) Max. 500.0 ppm
Heavy metals (as Pb) Max. 5 ppm
S-compounds (as SO₄) Max. 0.003 %
Ammonium (NH₄) Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR