Nickel (II) klorid heksahydrat ≥99.9995% (metals basis), Puratronic®

Leverandør: Thermo Fisher Scientific
Danger

43185.22 43185.14
43185.22EA 10075 NOK
43185.22 43185.14
Nickel (II) klorid heksahydrat ≥99.9995% (metals basis), Puratronic®
Nickel (II) klorid heksahydrat

Purity: 99.9995% CAS No.: 7791-20-0 Molecular Formula: Cl2Ni.6H2O Formula Weight: 237.71 (129.60anhy) Physical Form: Crystalline Density: 1.92 Appearance: Green Odour: Odourless pH: 4.9 Melting Point: 140° -H2O Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149809

Formel: NiCl₂·6H₂O
Smeltepunkt: 140 °C (-H₂O)
Tetthet: 1,92
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149809
CAS-nummer: 7791-20-0
EINECS: 231-743-0
FN: 3288
ADR: 6.1,III
Merck Indeks: 14,6505

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR