VWR®, Homogenisator, Potter-Elvehjem
Leverandør: VWR Collection
432-0200EA 730 NOK
432-0200 432-0201 432-0202 432-0203 432-0204 432-0205 432-0206 432-0207 432-0208 432-0209 432-0210 432-0211 432-0212 432-0213 432-0214 432-0215 432-0216 432-0217
VWR®, Homogenisator, Potter-Elvehjem
Homogenisatorer Vevshomogenisatorer
Borosilicate glass tube, PTFE pestle with stainless steel shaft. Tissue grinders are used for controlled reduction of particle size and homogenisation of a variety of substances, especially biological material. The shearing forces generated by the movement of the rotating plunger in a precision bore tube cause size reduction, determining factors include clearance between the pestle head and tube, speed of rotation and viscosity of medium.
Grinding chamber clearance: 0,15 to 0,25 mm.
Skaftet i rustfritt stål har en diameter på 6,5 mm.
Bestillingsinformasjon: Plunger must be ordered separately.