Cooling system for Precellys® Evolution, Cryolys® Evolution
Leverandør: Bertin Instruments
P000671-CLYS2A
432-0346EA 0 NOK
432-0346
Cooling system for Precellys® Evolution, Cryolys® Evolution
Homogenisatorer Homogenisatorer, bordmodell
Cryolys® Evolution is the ultimate cooling system for homogenising thermo-sensitive molecules with the Precellys® Evolution homogeniser, ensuring controlled and stable cooling before, during and after the homogenisation protocol. The Cryolys® Evolution prevents the increase of temperature during lysing and enhances the efficiency of molecular extraction, resulting in premium quality analysis, maintaining the desired temperature between 0 and 10 °C before and during homogenisation.
  • Very efficient cooling: Target temperature reached in less than 2 minutes