Zinkklorid 97.0-100.5%, granular USP, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

4320-07EA 23750 NOK
4320-07
Zinkklorid 97.0-100.5%, granular USP, J.T.Baker®
Zinkklorid
Formel: ZnCl₂
MW: 136.3 g/mol
Kokepunkt: 732 °C (1013 hPa)
Smeltepunkt: 283 °C
Tetthet: 2.91 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011295
CAS-nummer: 7646-85-7
EINECS: 231-592-0
FN: 2331
ADR: 8 III
Merck Indeks: 13,10185

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product
Meets U.S.P Requirements
Store at 2 to 25 °C
Preserve in Tight Containers Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical = 0.03 %
Identification Passes Test
Oxychloride Passes Test
Sulfate (SO₄) ≤0.03 %
Alkalies and Alkaline Earths ≤1.0 %
Ammonium Salts Passes Test
Lead (Pb) ≤0.005 %
Assay (ZnCl₂) 97.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR