Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Leverandør: Thermo Fisher Scientific

43296.09
43296.09EA 2550 NOK
43296.09
Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Magnesium

Purity: 99.95% CAS No.: 7439-95-4 Physical Form: ≈0.04g/slug Odour: Odourless MDL No.: MFCD00085308

Formel: Mg
Kokepunkt: 1107 °C (1013 hPa)
Smeltepunkt: 651 °C
Tetthet: 1,74 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085308
CAS-nummer: 7439-95-4
EINECS: 231-104-6
FN: 1869
ADR: 4.1,III
Merck Indeks: 13,05675

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR