Zirkonium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 0.25 mm (0.01 in)

Leverandør: Thermo Fisher Scientific

43334.G6
43334.G6EA 3350 NOK
43334.G6
Zirkonium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 0.25 mm (0.01 in)
Zirconium

Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈0.32g/m Odour: Odourless MDL No.: MFCD00011303

Formel: Zr
MW: 91.22 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011303
CAS-nummer: 7440-67-7
EINECS: 231-176-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR