Ruthenium(IV) oxide hydrate ≥99.99% (metals basis) Ru 54-60%, Premion®

Leverandør: Thermo Fisher Scientific
Warning

43403.14 43403.03 43403.06
43403.14EA 32075 NOK
43403.14 43403.03 43403.06
Ruthenium(IV) oxide hydrate ≥99.99% (metals basis) Ru 54-60%, Premion®
Ruthenium(IV) oxide hydrate

Purity: 99.99% CAS No.: 32740-79-7 Molecular Formula: RuO2.xH2O Formula Weight: 133.07(anhy) Physical Form: Powder Appearance: Black Melting Point: 1200° subl. UN No.: 1479 MDL No.: MFCD00149846

Formel: O₂Ru·H₂O
Smeltepunkt: 1200 °C (subl.)
Tetthet: 6,97 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149846
CAS-nummer: 32740-79-7
EINECS: 234-840-6
FN: 1479
ADR: ,

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR