Aluminium ≥99.9995% (metals basis), folie, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)

Leverandør: Thermo Fisher Scientific

43424.GH 43424.HP 43424.FI
43424.GHEA 2450 NOK
43424.GH 43424.HP 43424.FI
Aluminium ≥99.9995% (metals basis), folie, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)
Aluminium

Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈1.7g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029

Formel: Al
Kokepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Tetthet: 2,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Indeks: 13,00321

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR