Antimon(III)klorid, ultra dry ≥99.999% (metals basis)

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Antimony trichloride

Ampuled under argon

43449.22 43449.14
43449.22EA 11300 NOK
43449.22 43449.14
Antimon(III)klorid, ultra dry ≥99.999% (metals basis)
Antimon(III)klorid

Purity: 99.999% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Physical Form: Crystals, Ampouled under argon Density: 3.14 Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formel: SbCl₃
MW: 228.12 g/mol
Kokepunkt: 223.5 °C
Smeltepunkt: 73.4 °C
Tetthet: 3.14
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011212
CAS-nummer: 10025-91-9
EINECS: 233-047-2
FN: 1733
ADR: 8 II
Merck Indeks: 14,707

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR