5,8,11,14-Eicosatetraynoic acid, Millipore®

Leverandør: Merck Millipore Calbiochem

434741-20
434741-20EA 7850 NOK
434741-20
5,8,11,14-Eicosatetraynoic acid, Millipore®
5,8,11,14-Eicosatetraynoic acid
Formel: C₂₀H₂₄O₂
MDL nummer: MFCD00036967
CAS-nummer: 1191-85-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR