O-Ethylhydroxylamine hydrochloride 99.0-101.0% (by argentometric titration), LiChropur™ for GC derivatisation, Supelco®

Leverandør: Merck

Synonymer: Ethoxyamine hydrochloride

43504-10G
43504-10GEA 4790 NOK
43504-10G
O-Ethylhydroxylamine hydrochloride 99.0-101.0% (by argentometric titration), LiChropur™ for GC derivatisation, Supelco®
O-Ethylhydroxylamine hydrochloride
Formel: C₂H₈ClNO
MDL nummer: MFCD00012956
CAS-nummer: 3332-29-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c163faaf10efb5e81c530b9bb2922d84e7b7e17d&vid=bb9c65d6ff2db80a3abb50d9b30c50273c840be0&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR