Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal®

Leverandør: Thermo Fisher Scientific
Danger

435991000
435991000.EA 2575 NOK
435991000.
Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal®
Tetrabutylammonium dihydrogen phosphate
Formel: C₁₆H₃₈NO₄P
Kokepunkt: 81…82 °C (760 mmHg)
Smeltepunkt: –48 °C
Tetthet: 0,828 g/cm³
Flash punkt: 2 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064526
CAS-nummer: 5574-97-0
FN: 1648
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to light yellow Solution
Titration with NaOH 0.38 to 0.45 M
Melting point 215 to 220 °C
HPLC ≥96.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR