Etanol absolute ≥99.8%, Electran for molekylærbiologi

Leverandør: VWR Chemicals
Danger

437433TEA 1025 NOK
437433T 437433TDP 437435L
Etanol absolute ≥99.8%, Electran for molekylærbiologi
Etanol

Bestillingsinformasjon: WARNING : This alcohol is regulated. Please contact VWR.

Formel: C₂H₅OH
Kokepunkt: 78,3 °C (1013 hPa)
Smeltepunkt: –117 °C
Tetthet: 0,7895 g/cm³ (20 °C)
Flash punkt: 12 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
FN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Indeks: 12,03806

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.8 %
Appearance Clear colourless liquid
DNases (exo- and endonucleases) Not detected
RNases Not detected
Proteases Not detected
Acidity or alkalinity Max. 0.0002 meq/g
Heavy metals (as Pb) Max. 1 ppm
Non-volatile residue Max. 0.0010 %
Water Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR