Multi-element standard solution 6 for ICP, TraceCERT®
Leverandør: Merck
43843-100ML
43843-100MLEA 1675 NOK
43843-100ML
Multi-element standard solution 6 for ICP, TraceCERT®
Standarder Kationstandarder Multi-Element Standards
This standard solution is intended for use as a certified reference material for ICP/AAS.
Multi element standards for Al, Sb, Ba, Pb, B, Ca, Cd, Cr, Co, Fe, K, Cu, Li, Mg, Mn, Mo, Na, Ni, P, Si, Ti, V and Zn at 100 mg/ml in 5% nitric acid and ≤0,1% hydrofluoric acid.
Sertifiseringer: Accredited to ISO 17034 and ISO/IEC 17025.