Bismuth standard dissolved in oil, 5.000 µg/g Bi in mineral oil Specpure® til wear metal analysis
Leverandør: Thermo Fisher Scientific
43865.1Y 43865.18
43865.1YEA 920 NOK
43865.1Y 43865.18
Bismuth standard dissolved in oil, 5.000 µg/g Bi in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Bismuth Standards
Physical form: liquid
MDL No.: MFCD00134033
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
MDL No. MFCD00134033