Sulphur standard dissolved in oil, 1.000 µg/g S in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

43869.1Y 43869.18
43869.1YEA 680 NOK
43869.1Y 43869.18
Sulphur standard dissolved in oil, 1.000 µg/g S in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Sulphur Standards

Physical form: liquid
MDL No.: MFCD00242296

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR