Dimetylsulfoksid, vannfri ≥99.8%

Leverandør: Thermo Fisher Scientific
Warning


Packaged under argon in resealable ChemSeal™ bottles.

43998.K2 43998.M1
43998.K2EA 3650 NOK
43998.K2 43998.M1
Dimetylsulfoksid, vannfri ≥99.8%
Dimetylsulfoksid

Purity: 99.8+% CAS No.: 67-68-5 Molecular Formula: (CH3)2SO Formula Weight: 78.13 Flash Point: 87°(189°F) Physical Form: Liquid Density: 1.101 Refractive Index: 1.4790 Sensitivity: Hygroscopic Odour: Mild Melting Point: 18.4° Boiling Point: 189° Beilstein Reference: 506008 Merck Reference: 14,3259 MDL No.: MFCD00002089

Formel: (CH₃)₂SO
Kokepunkt: 189 °C
Tetthet: 1,101
Flash punkt: 87 °C (189 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Indeks: 14,3259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR