Tetrabutylammonium dihydrogen phosphate 1 M in ethanol, AcroSeal®

Leverandør: Thermo Fisher Scientific
Danger

440061000
440061000.EA 1525 NOK
440061000.
Tetrabutylammonium dihydrogen phosphate 1 M in ethanol, AcroSeal®
Tetrabutylammonium dihydrogen phosphate
Formel: C₁₆H₃₈NO₄P
Tetthet: 0,895 g/cm³
Flash punkt: 14 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064526
CAS-nummer: 5574-97-0
FN: 1170
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Solution
Titration with NaOH 0.95 to 1.10 M

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR