L(+)-Askorbinsyre ≥99.0%, AR® ACS, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

Synonymer: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

4407-04EA 5425 NOK
4407-04
L(+)-Askorbinsyre ≥99.0%, AR® ACS, Macron Fine Chemicals™
L(+)-Askorbinsyre

Crystals.

Formel: C₆H₈O₆
MW: 176.13 g/mol
Kokepunkt: 553
Smeltepunkt: 190…192 °C
Tetthet: 1.694 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064328
CAS-nummer: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Indeks: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use Passes Test
ACS - Assay (C₆H₈O₆) ≥99.0 %
ACS - Appearance (colorless or white to slightly yellow crystals) Passes Test
Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Residue on Ignition ≤0.10 %
Specific Rotation, [Å]²⁵^D(c=10 in water)(+) 20.5 - 21.5 Degree

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR