Silver conductive adhesive paste

Leverandør: Thermo Fisher Scientific
Warning

44075.18
44075.18EA 8625 NOK
44075.18
Silver conductive adhesive paste
Silver conductive adhesive paste

Physical Form: Nominal electrical resistivity: 1x10-4ohm-cm Appearance: Silver paste Odour: Aromatic MDL No.: MFCD01868500

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01868500
EINECS: 232-033-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR