Mechanical overhead stirrer, RW 28 digital
Leverandør: IKA
9023202 5040000 5040002
441-0338EA 33375 NOK
441-0338 441-0329 441-0330
Mechanical overhead stirrer, RW 28 digital
Omrørere Omrører
Powerful, mechanically controlled laboratory stirrer, designed for highly viscous applications for quantities up to 80 L. Units have a robust, ergonomic design with constant power drive, suitable for intensive mixing for use in laboratories and pilot plants. The stirrer is equipped with protection to avoid overheating the motor by means of a self-locking temperature limiter. Additionally, to secure bowls, a stirring shaft protector and clamping fixture are optionally available.
  • Digital speed display
  • Infinitely adjustable speed in two speed ranges
  • Push