HEPES ≥99.5%

Leverandør: VWR Chemicals

Synonymer: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

441476LEA 11300 NOK
441476L
HEPES ≥99.5%
HEPES

Hepes is a widely used buffer in biological studies. In cell culture media, it is employed as a substitute for the bicarbonate buffer at a concentration of 25 mM or as a supplement to the bicarbonate buffer (concentration 10 to 15 mM).

Formel: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Smeltepunkt: 210…215 °C
Tetthet: 1.448 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006158
CAS-nummer: 7365-45-9
EINECS: 230-907-9
Merck Indeks: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.5 %
Cl (Chloride) Max. 0.05 %
SO4 (Sulphate) Max. 0.05 %
Heavy metals (as Pb) Max. 10 ppm
pH (25°C; 1 %) 4.7 - 6.0
Water Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR