Platina nominally 60% on carbon black

Supplier: Thermo Fisher Scientific

Synonyms: Platinium

44171.18
44171.18EA 0 NOK
44171.18
Platina nominally 60% on carbon black
Platina
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR