Platina ≥99.9% (metals basis), wire, Ø 0.01 mm (0.0004 in)

Leverandør: Thermo Fisher Scientific

Synonymer: Platinium

44196.G1
44196.G1EA 11550 NOK
44196.G1
Platina ≥99.9% (metals basis), wire, Ø 0.01 mm (0.0004 in)
Platina

Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈1.68mg/m, Temper: hard Odour: Odourless MDL No.: MFCD00011179

Formel: Pt
MW: 195.08 g/mol
Kokepunkt: 3827
Smeltepunkt: 1769
Tetthet: 21.447 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011179
CAS-nummer: 7440-06-4
EINECS: 231-116-1
Merck Indeks: 12,07689

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR