2-Dodecylcyclobutanone, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

44197-5MG
44197-5MGEA 2350 NOK
44197-5MG
2-Dodecylcyclobutanone, Supelco®
2-Dodecylcyclobutanone
Formel: C₁₆H₃₀O
MW: 238.41 g/mol
Kokepunkt: 322
Smeltepunkt: 28…31 °C
Lagringstemperatur: Romtemperatur
CAS-nummer: 35493-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=761c2ee132959a4be9f194415e8c7b20061a50fd&vid=30b9d5963d66a1e64ca009133820f9ea02a76f75&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR