VWR®, Magnetic Stirrer, Lab Disc
Supplier: VWR Collection
442-0883EA 4850 NOK
442-0883
VWR®, Magnetic Stirrer, Lab Disc
Stirrers Magnetic Stirrers
Extremely flat, compact magnetic stirrer with no moving parts. Reverses direction of rotation every 30 seconds to ensure optimum mixing.
  • Selectable reversible rotation direction
  • Wear-free drive
  • Set-up plate and casing made of chemically resistant materials
  • Slip proof, safe stand
IP protection class according to DIN EN 60529: IP 65
Certifications: IP protection class according to DIN EN 60529: IP 65
Delivery information: Supplied with two beakers (150 ml) and three magnetic stirring bars (10 mm, 20 mm, 30 mm).
Order Now