Multi-position magnetic stirrers, RO 5/10/15
Leverandør: IKA
3692500 3690500 3691000
442-0921EA 35025 NOK
442-0921 442-0919 442-0920
Multi-position magnetic stirrers, RO 5/10/15
Omrørere Magnetrørere, flerposisjons
Compact digital magnetic stirrers with 5, 10 or 15 stirring positions, designed for synchronous stirring and suitable for continuous operation. The wear-free magnetic coils provide noiseless and consistent stirring at all positions. Speed remains constant, even when load changes. The design allows easy cleaning and protects the equipment from liquid spills.
  • Adjustable and reversible operation
  • Error code display
  • Easy operation with touch keypad
  • Speed is adjustable in increments of 10 min⁻¹
IP protection class according to DIN EN 60529: IP 40