Magnetic stirrer, myPlate
Leverandør: IKA
25005156
442-1262EA 5350 NOK
442-1262
Magnetic stirrer, myPlate
Omrørere Magnetrørere
The myPlate magnetic stirrer has similar technical features to the popular colour squid and big squid magnetic stirrers together with the option of creative individualisation. The interchangeable personal frame allows you to design the set-up plate of the myPlate by yourself.
  • High chemical resistance
  • Electronically controlled motor
  • Interchangeable frame for customisation
  • Proven high functionality
  • Digital speed indicator
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
The stirrer is supplied with a decorative imageMore Product Information